Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCTACTTTTGTAATGTCTTGAGA[A/G]AGCGTCGTCAGACTGTTGTTCAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609861 | ||||||||||||||||||||
Literature Links: |
IKBIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IKBIP - IKBKB interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153687.3 | 682 | Intron | NP_710154.1 | |||
NM_201612.2 | 682 | Silent Mutation | CTC,CTT | L,L 150 | NP_963906.1 | |
NM_201613.2 | 682 | UTR 3 | NP_963907.1 |