Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCGGCGCCTTTAATTGGGGCTC[C/T]GGCTAACTCTGAGGACACGCATTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612456 MIM: 600778 MIM: 602927 | ||||||||||||||||||||
Literature Links: |
APOLD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOLD1 - apolipoprotein L domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDKN1B - cyclin dependent kinase inhibitor 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004064.4 | 935 | Missense Mutation | CCG,CTG | P,L 122 | NP_004055.1 |
GPR19 - G protein-coupled receptor 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006143.2 | 935 | Intron | NP_006134.1 |