Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACCGTGGAGCAGCTACTTTAA[C/T]AGTGTTGCCAAATTTGGAGTAATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600935 | ||||||||||||||||||||
Literature Links: |
KCNJ8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNJ8 - potassium voltage-gated channel subfamily J member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004982.3 | 1351 | Missense Mutation | ATT,GTT | I,V 346 | NP_004973.1 | |
XM_005253358.4 | 1351 | Missense Mutation | ATT,GTT | I,V 346 | XP_005253415.1 | |
XM_017019283.1 | 1351 | Missense Mutation | ATT,GTT | I,V 346 | XP_016874772.1 | |
XM_017019284.1 | 1351 | Missense Mutation | ATT,GTT | I,V 346 | XP_016874773.1 |