Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGAATGAGCACAGCCGCTGGC[A/G]GCCCTACACGGCCACCGTGTGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602582 | ||||||||||||||||||||
Literature Links: |
DTX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DTX1 - deltex E3 ubiquitin ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004416.2 | 1091 | Missense Mutation | CAG,CGG | Q,R 38 | NP_004407.2 | |
XM_011538009.2 | 1091 | Missense Mutation | CAG,CGG | Q,R 38 | XP_011536311.1 |