Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCGTAACTGGAGGAGGCCACCT[C/T]GGTCTTCCAGACAGCCTCCATCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM186B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM186B - family with sequence similarity 186 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032130.2 | 2734 | Missense Mutation | AAG,GAG | K,E 858 | NP_115506.1 | |
XM_006719625.2 | 2734 | Missense Mutation | AAG,GAG | K,E 858 | XP_006719688.1 | |
XM_006719626.2 | 2734 | Intron | XP_006719689.1 | |||
XM_006719627.3 | 2734 | Intron | XP_006719690.1 | |||
XM_011538796.2 | 2734 | Missense Mutation | AAG,GAG | K,E 858 | XP_011537098.1 | |
XM_011538797.2 | 2734 | Intron | XP_011537099.1 | |||
XM_017020008.1 | 2734 | Missense Mutation | AAG,GAG | K,E 768 | XP_016875497.1 |
LOC100422296 - Parkinsonism associated deglycase pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |