Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATACTTACTTACAATAACTACCA[C/T]GATGATGGCACAGATGGCTCCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607081 MIM: 185880 | ||||||||||||||||||||
Literature Links: |
TAPBPL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TAPBPL - TAP binding protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018009.4 | 693 | Intron | NP_060479.3 | |||
XM_005253700.3 | 693 | Intron | XP_005253757.1 | |||
XM_017019545.1 | 693 | Intron | XP_016875034.1 | |||
XM_017019546.1 | 693 | Intron | XP_016875035.1 |
VAMP1 - vesicle associated membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297438.1 | 693 | Missense Mutation | ATG,GTG | M,V 109 | NP_001284367.1 | |
NM_014231.4 | 693 | Missense Mutation | ATG,GTG | M,V 109 | NP_055046.1 | |
NM_016830.3 | 693 | Missense Mutation | ATG,GTG | M,V 109 | NP_058439.1 | |
NM_199245.2 | 693 | Missense Mutation | ATG,GTG | M,V 109 | NP_954740.1 |