Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCAGGAGGAGCCTGGGACGCC[A/G]CCCTCCTCGCCGCTGAGTGCCGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610602 MIM: 191525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALKBH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALKBH2 - alkB homolog 2, alpha-ketoglutarate dependent dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNG - uracil DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003362.3 | 269 | Silent Mutation | CCA,CCG | P,P 52 | NP_003353.1 | |
NM_080911.2 | 269 | Silent Mutation | CCA,CCG | P,P 61 | NP_550433.1 |