Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCTTTGTCTTTTGTCCTCAGC[C/T]TCAGACTATGGCATGAAACTGCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613802 | ||||||||||||||||||||
Literature Links: |
MLEC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MLEC - malectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303627.1 | 410 | UTR 5 | NP_001290556.1 | |||
NM_001303628.1 | 410 | Silent Mutation | GCC,GCT | A,A 79 | NP_001290557.1 | |
NM_014730.3 | 410 | Silent Mutation | GCC,GCT | A,A 79 | NP_055545.1 | |
XM_011539031.1 | 410 | UTR 5 | XP_011537333.1 | |||
XM_011539032.1 | 410 | UTR 5 | XP_011537334.1 |