Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGGAGTTCACAGTGGGAGACAA[A/C]CCTGTTTCAAACTTCCGGATGATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606885 | ||||||||||||||||||||
Literature Links: |
ACADS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACADS - acyl-CoA dehydrogenase, C-2 to C-3 short chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4700 - microRNA 4700 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC119B - unc-119 lipid binding chaperone B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080533.2 | 535 | Silent Mutation | AAA,AAC | K,N 165 | NP_001074002.1 |