Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTCCTCCAGTTATTTAAATCTG[A/G]TGAAGAAATAAAAGGAAAACAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179838 MIM: 610034 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLIP1 - CAP-Gly domain containing linker protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001247997.1 | 5209 | UTR 3 | NP_001234926.1 | |||
NM_002956.2 | 5209 | UTR 3 | NP_002947.1 | |||
NM_198240.1 | 5209 | UTR 3 | NP_937883.1 | |||
XM_005253593.2 | 5209 | UTR 3 | XP_005253650.1 | |||
XM_006719552.3 | 5209 | UTR 3 | XP_006719615.1 | |||
XM_017019786.1 | 5209 | UTR 3 | XP_016875275.1 | |||
XM_017019787.1 | 5209 | UTR 3 | XP_016875276.1 | |||
XM_017019788.1 | 5209 | UTR 3 | XP_016875277.1 | |||
XM_017019789.1 | 5209 | UTR 3 | XP_016875278.1 | |||
XM_017019790.1 | 5209 | UTR 3 | XP_016875279.1 | |||
XM_017019791.1 | 5209 | UTR 3 | XP_016875280.1 | |||
XM_017019792.1 | 5209 | UTR 3 | XP_016875281.1 |
VPS33A - VPS33A, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |