Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCTGGACAATGAGTCCTTTTACA[C/G]CAAAGCGTCTGCAGGCAAATGCGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601527 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALX1 - ALX homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006982.2 | 129 | Missense Mutation | ACC,AGC | T,S 42 | NP_008913.2 | |
XM_011538782.2 | 129 | Intron | XP_011537084.1 |
LRRIQ1 - leucine rich repeats and IQ motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |