Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGCAGGCACCAGGGGTCCCACC[A/G]ATGGACACACCTTGGAGGCAGCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611181 MIM: 604986 | ||||||||||||||||||||
Literature Links: |
ACAD10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACAD10 - acyl-CoA dehydrogenase family member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136538.1 | 298 | Missense Mutation | CAA,CGA | Q,R 33 | NP_001130010.1 | |
NM_025247.5 | 298 | Missense Mutation | CAA,CGA | Q,R 33 | NP_079523.3 |
BRAP - BRCA1 associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |