Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACCACCCTCAGAGGCCGATCTG[A/C]CGCTGACCTGGTTGTCTTCCTCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164350 MIM: 612159 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OAS1 - 2'-5'-oligoadenylate synthetase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032409.2 | 314 | Missense Mutation | GAC,GCC | D,A 75 | NP_001027581.1 | |
NM_001320151.1 | 314 | Missense Mutation | GAC,GCC | D,A 75 | NP_001307080.1 | |
NM_002534.3 | 314 | Missense Mutation | GAC,GCC | D,A 75 | NP_002525.2 | |
NM_016816.3 | 314 | Missense Mutation | GAC,GCC | D,A 75 | NP_058132.2 | |
XM_006719434.1 | 314 | Missense Mutation | GAC,GCC | D,A 75 | XP_006719497.1 | |
XM_011538413.1 | 314 | Missense Mutation | GAC,GCC | D,A 75 | XP_011536715.1 | |
XM_017019361.1 | 314 | Missense Mutation | GAC,GCC | D,A 75 | XP_016874850.1 | |
XM_017019362.1 | 314 | UTR 5 | XP_016874851.1 |
RPH3A - rabphilin 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |