Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCCTGATCTCAGTGAGGTCCTC[C/T]TTGGTGAACACAAAGCCCACATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605614 MIM: 180510 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCN1 - GCN1, eIF2 alpha kinase activator homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PXN-AS1 - PXN antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPLP0 - ribosomal protein lateral stalk subunit P0 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002.3 | Intron | NP_000993.1 | ||||
NM_053275.3 | Intron | NP_444505.1 |