Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGAAGACCAAGATGGTGGCTG[C/T]GTTGTTGAGCTGGTTGAGTCGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612220 MIM: 607297 | ||||||||||||||||||||
Literature Links: |
B4GALNT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B4GALNT3 - beta-1,4-N-acetyl-galactosaminyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NINJ2 - ninjurin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294345.1 | 700 | Missense Mutation | NP_001281274.1 | |||
NM_001294346.1 | 700 | Missense Mutation | NP_001281275.1 | |||
NM_016533.5 | 700 | Missense Mutation | NP_057617.2 |