Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAATTGAATGAGACTCTCTAGGG[G/T]ATGAGACTCAGGCATCAGCATTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138252 | ||||||||||||||||||||
Literature Links: |
GRIN2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRIN2B - glutamate ionotropic receptor NMDA type subunit 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000834.3 | 30462 | Intron | NP_000825.2 | |||
XM_005253351.3 | 30462 | UTR 3 | XP_005253408.1 | |||
XM_011520628.2 | 30462 | UTR 3 | XP_011518930.1 | |||
XM_011520629.2 | 30462 | UTR 3 | XP_011518931.1 | |||
XM_017019219.1 | 30462 | UTR 3 | XP_016874708.1 |