Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGTGTCTGGGCTTCCGCAGCT[G/T]CTTCTTCTTGAAGTAAGCATCAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603703 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPL6 - ribosomal protein L6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000970.4 | 940 | Intron | NP_000961.2 | |||
NM_001024662.2 | 940 | Intron | NP_001019833.1 | |||
NM_001320137.1 | 940 | Intron | NP_001307066.1 | |||
NM_001320138.1 | 940 | Missense Mutation | AAG,CAG | K,Q 221 | NP_001307067.1 | |
NM_001320139.1 | 940 | Intron | NP_001307068.1 | |||
NM_001320140.1 | 940 | Missense Mutation | AAG,CAG | K,Q 221 | NP_001307069.1 | |
NM_001320141.1 | 940 | Missense Mutation | AAG,CAG | K,Q 221 | NP_001307070.1 | |
NM_001320142.1 | 940 | Intron | NP_001307071.1 | |||
XM_017019781.1 | 940 | Missense Mutation | AAG,CAG | K,Q 221 | XP_016875270.1 | |
XM_017019782.1 | 940 | Intron | XP_016875271.1 |