Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGAGACTCGGGGCTCCGAGGA[C/T]GAGACGGACGACGATGGGGAAGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142410 | ||||||||||||||||||||
Literature Links: |
HNF1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNF1A - HNF1 homeobox A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000545.6 | 417 | Silent Mutation | GAC,GAT | D,D 72 | NP_000536.5 | |
NM_001306179.1 | 417 | Silent Mutation | GAC,GAT | D,D 72 | NP_001293108.1 | |
XM_005253931.3 | 417 | Silent Mutation | GAC,GAT | D,D 72 | XP_005253988.1 |
HNF1A-AS1 - HNF1A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |