Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGATCACTAGCCAGGAGATCTC[C/T]GCTGCGCATTCCTACAACCCCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KLHL42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL42 - kelch like family member 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020782.1 | 1181 | Silent Mutation | TCC,TCT | S,S 269 | NP_065833.1 | |
XM_017019698.1 | 1181 | Silent Mutation | TCC,TCT | S,S 269 | XP_016875187.1 |
MANSC4 - MANSC domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |