Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCTTTTATTCAAGTAAAAAAAT[A/G]TTCTTCCTCTTGTGATTTTGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603541 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC38 - coiled-coil domain containing 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182496.2 | 1705 | Missense Mutation | CAT,TAT | H,Y 560 | NP_872302.2 | |
XM_006719229.2 | 1705 | Missense Mutation | CAT,TAT | H,Y 396 | XP_006719292.1 | |
XM_011537883.2 | 1705 | Missense Mutation | CAT,TAT | H,Y 560 | XP_011536185.1 | |
XM_011537884.2 | 1705 | Missense Mutation | CAT,TAT | H,Y 520 | XP_011536186.1 | |
XM_011537886.2 | 1705 | Missense Mutation | CAT,TAT | H,Y 520 | XP_011536188.1 | |
XM_011537887.2 | 1705 | Missense Mutation | CAT,TAT | H,Y 520 | XP_011536189.1 | |
XM_011537888.2 | 1705 | Missense Mutation | CAT,TAT | H,Y 343 | XP_011536190.1 | |
XM_011537889.1 | 1705 | Intron | XP_011536191.1 |
LOC105369921 - uncharacterized LOC105369921 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRPF - small nuclear ribonucleoprotein polypeptide F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003095.2 | 1705 | Intron | NP_003086.1 |