Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTATTACAGGTGCAGATGAAGA[C/G]GCTCACTTTTTTCTGGTCTTTAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606782 MIM: 612252 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC1A - C-type lectin domain family 1 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297748.1 | 1772 | UTR 3 | NP_001284677.1 | |||
NM_001297749.1 | 1772 | UTR 3 | NP_001284678.1 | |||
NM_001297750.1 | 1772 | UTR 3 | NP_001284679.1 | |||
NM_001297751.1 | 1772 | UTR 3 | NP_001284680.1 | |||
NM_016511.3 | 1772 | UTR 3 | NP_057595.2 | |||
XM_011520687.2 | 1772 | UTR 3 | XP_011518989.1 |
CLEC9A - C-type lectin domain family 9 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |