Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCTGAGTAAAGAAGAAGTAAAA[C/T]GCCTCATTGCTGAGGCAAAGGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PWP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PWP1 - PWP1 homolog, endonuclein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317962.1 | 124 | UTR 5 | NP_001304891.1 | |||
NM_001317963.1 | 124 | UTR 5 | NP_001304892.1 | |||
NM_007062.2 | 124 | Missense Mutation | CGC,TGC | R,C 34 | NP_008993.1 |