Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCGCGCTGCTGGCCGCCTGCTGC[G/T]GGGGGGAAGCGCTCGTCCAGATTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE3A - phosphodiesterase 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000921.4 | 545 | Missense Mutation | GGG,TGG | G,W 169 | NP_000912.3 | |
NM_001244683.1 | 545 | Intron | NP_001231612.1 | |||
XM_017019420.1 | 545 | Missense Mutation | GGG,TGG | G,W 169 | XP_016874909.1 | |
XM_017019421.1 | 545 | Intron | XP_016874910.1 |