Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTCTCAAAGCGACTGTATTCC[C/T]GGCGGTCATAGATTTCCACCGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147559 | ||||||||||||||||||||
Literature Links: |
ITGB7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGB7 - integrin subunit beta 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000889.2 | 2499 | Missense Mutation | CAG,CGG | Q,R 756 | NP_000880.1 | |
XM_005268851.3 | 2499 | Missense Mutation | CAG,CGG | Q,R 756 | XP_005268908.1 | |
XM_005268852.4 | 2499 | Missense Mutation | CAG,CGG | Q,R 756 | XP_005268909.1 | |
XM_006719376.3 | 2499 | Missense Mutation | CAG,CGG | Q,R 756 | XP_006719439.1 |
ZNF740 - zinc finger protein 740 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |