Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGAGGAGGAGGAAGAACACCA[A/G]GAATCATCATCCTCGCCGTAGAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608500 | ||||||||||||||||||||
Literature Links: |
PRICKLE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRICKLE1 - prickle planar cell polarity protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144881.1 | 2388 | Silent Mutation | NP_001138353.1 | |||
NM_001144882.1 | 2388 | Silent Mutation | NP_001138354.1 | |||
NM_001144883.1 | 2388 | Silent Mutation | NP_001138355.1 | |||
NM_153026.2 | 2388 | Silent Mutation | NP_694571.2 | |||
XM_011537946.1 | 2388 | Silent Mutation | XP_011536248.1 | |||
XM_011537947.2 | 2388 | Silent Mutation | XP_011536249.1 | |||
XM_017018838.1 | 2388 | Silent Mutation | XP_016874327.1 | |||
XM_017018839.1 | 2388 | Silent Mutation | XP_016874328.1 | |||
XM_017018840.1 | 2388 | Silent Mutation | XP_016874329.1 |