Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGGTTAAAGCTTTTAAGTGTTT[C/T]AAAGTTAATTCCAAGACTACAGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606200 MIM: 601599 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BHLHE41 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BHLHE41 - basic helix-loop-helix family member e41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030762.2 | 570 | Silent Mutation | TTA,TTG | L,L 93 | NP_110389.1 |
SSPN - sarcospan | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135823.1 | 570 | Intron | NP_001129295.1 | |||
NM_005086.4 | 570 | Intron | NP_005077.2 | |||
XM_011520853.2 | 570 | Intron | XP_011519155.1 | |||
XM_011520854.2 | 570 | Intron | XP_011519156.1 | |||
XM_011520855.2 | 570 | Intron | XP_011519157.1 |