Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCATCTTGAAATATGAGTTTGTC[A/G]AGGTGCTCTATGATCTCATCAATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609509 | ||||||||||||||||||||
Literature Links: |
IL31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL31 - interleukin 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014336.1 | 360 | Silent Mutation | CTC,CTT | L,L 111 | NP_001014358.1 |
LRRC43 - leucine rich repeat containing 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098519.1 | 360 | Intron | NP_001091989.1 | |||
NM_152759.4 | 360 | Intron | NP_689972.3 | |||
XM_017019124.1 | 360 | Intron | XP_016874613.1 | |||
XM_017019125.1 | 360 | Intron | XP_016874614.1 |