Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGGTAACGGTGTCCTTTGTTCT[C/T]CAAGTATGTGATGAAGGGAAGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609992 MIM: 615998 | ||||||||||||||||||||
Literature Links: |
POP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POP5 - POP5 homolog, ribonuclease P/MRP subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015918.3 | 294 | Missense Mutation | AAG,GAG | K,E 88 | NP_057002.2 | |
NM_198202.1 | 294 | Intron | NP_937845.1 | |||
XM_011538441.1 | 294 | Missense Mutation | AAG,GAG | K,E 88 | XP_011536743.1 |
RNF10 - ring finger protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |