Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCGACTCAGCCCGCTCCTCCC[C/T]TTCCTCCTGTGTTTTCTGACGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605864 MIM: 605219 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GNT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B3GNT4 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011538732.1 | 1063 | Intron | XP_011537034.1 |
DIABLO - diablo IAP-binding mitochondrial protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278302.1 | 1063 | Missense Mutation | AGG,GGG | R,G 127 | NP_001265231.1 | |
NM_001278303.1 | 1063 | Missense Mutation | AGG,GGG | R,G 151 | NP_001265232.1 | |
NM_001278304.1 | 1063 | Missense Mutation | AGG,GGG | R,G 171 | NP_001265233.1 | |
NM_001278342.1 | 1063 | Missense Mutation | AGG,GGG | R,G 180 | NP_001265271.1 | |
NM_019887.5 | 1063 | Missense Mutation | AGG,GGG | R,G 224 | NP_063940.1 | |
NM_138930.3 | 1063 | Missense Mutation | AGG,GGG | R,G 171 | NP_620308.1 |
LRRC43 - leucine rich repeat containing 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |