Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAACTCTGTGTTACTAGACTGGA[A/G]TGAACAAGAGGTAGAATAGTGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM263 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM263 - transmembrane protein 263 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319661.1 | Intron | NP_001306590.1 | ||||
NM_001319662.1 | Intron | NP_001306591.1 | ||||
NM_001319663.1 | Intron | NP_001306592.1 | ||||
NM_001319664.1 | Intron | NP_001306593.1 | ||||
NM_001319666.1 | Intron | NP_001306595.1 | ||||
NM_152261.3 | Intron | NP_689474.1 | ||||
XM_017020198.1 | Intron | XP_016875687.1 |