Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAATGGGCTGTAGTGCACTTTG[A/G]GCGAGGGCGGCCTGGGGACCGCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601517 MIM: 605093 | ||||||||||||||||||||
Literature Links: |
ATXN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATXN2 - ataxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310121.1 | 3289 | Silent Mutation | GCC,GCT | A,A 1042 | NP_001297050.1 | |
NM_001310123.1 | 3289 | Silent Mutation | GCC,GCT | A,A 975 | NP_001297052.1 | |
NM_002973.3 | 3289 | Silent Mutation | GCC,GCT | A,A 1282 | NP_002964.3 |
SH2B3 - SH2B adaptor protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |