Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATTCGCAATTTCTTCAACATAG[A/G]TTTCATCCTCTAAAAGTACCCATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616929 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTERF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTERF2 - mitochondrial transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033050.2 | 580 | Missense Mutation | NP_001028222.1 | |||
NM_025198.4 | 580 | Missense Mutation | NP_079474.2 | |||
XM_011538767.2 | 580 | Missense Mutation | XP_011537069.1 | |||
XM_017019985.1 | 580 | Missense Mutation | XP_016875474.1 | |||
XM_017019986.1 | 580 | Missense Mutation | XP_016875475.1 | |||
XM_017019987.1 | 580 | Missense Mutation | XP_016875476.1 |
TMEM263 - transmembrane protein 263 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |