Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAGTGTTGATCACACCTAGCCC[A/T]CACCACAGGAGGGGGAATGAAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608352 MIM: 608524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACRBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACRBP - acrosin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ING4 - inhibitor of growth family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127582.1 | 1179 | UTR 3 | NP_001121054.1 | |||
NM_001127583.1 | 1179 | UTR 3 | NP_001121055.1 | |||
NM_001127584.1 | 1179 | UTR 3 | NP_001121056.1 | |||
NM_001127585.1 | 1179 | UTR 3 | NP_001121057.1 | |||
NM_001127586.1 | 1179 | UTR 3 | NP_001121058.1 | |||
NM_016162.3 | 1179 | UTR 3 | NP_057246.2 | |||
XM_005253698.3 | 1179 | UTR 3 | XP_005253755.1 | |||
XM_011520964.2 | 1179 | UTR 3 | XP_011519266.1 | |||
XM_011520965.2 | 1179 | UTR 3 | XP_011519267.1 | |||
XM_017019392.1 | 1179 | UTR 3 | XP_016874881.1 | |||
XM_017019393.1 | 1179 | UTR 3 | XP_016874882.1 |