Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTCAAATGTCACTTTATGAACG[A/G]CAAGCAGTGCAGGTGAGACTCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602978 | ||||||||||||||||||||
Literature Links: |
PHC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHC1 - polyhomeotic homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004426.2 | 602 | Intron | NP_004417.2 | |||
XM_005253334.2 | 602 | Intron | XP_005253391.1 | |||
XM_011520599.2 | 602 | Intron | XP_011518901.1 | |||
XM_011520600.2 | 602 | Intron | XP_011518902.1 | |||
XM_011520603.2 | 602 | Intron | XP_011518905.1 | |||
XM_017018955.1 | 602 | Silent Mutation | CGA,CGG | R,R 34 | XP_016874444.1 | |
XM_017018956.1 | 602 | Intron | XP_016874445.1 | |||
XM_017018957.1 | 602 | Intron | XP_016874446.1 | |||
XM_017018958.1 | 602 | Intron | XP_016874447.1 |