Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTTGAGCAGCAAGAAGGGGATG[C/T]AGCTCAGGGCAAAGCAGAGGAACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606383 MIM: 609124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR84 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR84 - G protein-coupled receptor 84 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020370.2 | 1784 | Missense Mutation | TAC,TGC | Y,C 332 | NP_065103.1 | |
XM_011538495.2 | 1784 | Missense Mutation | TAC,TGC | Y,C 332 | XP_011536797.1 |
LOC102724050 - uncharacterized LOC102724050 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF385A - zinc finger protein 385A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |