Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGTTCAGCTGCTGTCCAGACC[C/T]GGATCGGCAACAGTGCCGCCTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609858 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETNK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETNK1 - ethanolamine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039481.1 | 107 | Missense Mutation | CGG,TGG | R,W 30 | NP_001034570.1 | |
NM_018638.4 | 107 | Missense Mutation | CGG,TGG | R,W 30 | NP_061108.2 | |
XM_005253420.4 | 107 | Missense Mutation | CGG,TGG | R,W 30 | XP_005253477.1 | |
XM_017019580.1 | 107 | Missense Mutation | CGG,TGG | R,W 30 | XP_016875069.1 | |
XM_017019581.1 | 107 | Missense Mutation | CGG,TGG | R,W 30 | XP_016875070.1 |
LOC105369691 - uncharacterized LOC105369691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |