Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCCACCAGCAGCGGGGCAAGA[A/G]ACTGGTTTTCCTCTGGGTTGGGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601657 | ||||||||||||||||||||
Literature Links: |
CCER1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCER1 - coiled-coil glutamate rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152638.3 | 1243 | Missense Mutation | TCT,TTT | S,F 270 | NP_689851.1 |
EPYC - epiphycan | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC00615 - long intergenic non-protein coding RNA 615 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |