Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGAACCACAGCCGCTGCCGCCGC[C/T]GCCGCCGCCATTGGTCCGTAGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603328 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSI1 - musashi RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002442.3 | 1588 | Silent Mutation | GCA,GCG | A,A 276 | NP_002433.1 | |
XM_006719403.3 | 1588 | Silent Mutation | GCA,GCG | A,A 276 | XP_006719466.1 | |
XM_006719404.3 | 1588 | Silent Mutation | GCA,GCG | A,A 138 | XP_006719467.1 | |
XM_011538361.2 | 1588 | Silent Mutation | GCA,GCG | A,A 276 | XP_011536663.1 | |
XM_011538362.2 | 1588 | Silent Mutation | GCA,GCG | A,A 257 | XP_011536664.1 | |
XM_011538363.2 | 1588 | Silent Mutation | GCA,GCG | A,A 257 | XP_011536665.1 | |
XM_011538364.2 | 1588 | Silent Mutation | GCA,GCG | A,A 176 | XP_011536666.1 | |
XM_011538365.2 | 1588 | Silent Mutation | GCA,GCG | A,A 173 | XP_011536667.1 | |
XM_011538366.2 | 1588 | Silent Mutation | GCA,GCG | A,A 173 | XP_011536668.1 | |
XM_011538368.2 | 1588 | Silent Mutation | GCA,GCG | A,A 138 | XP_011536670.1 | |
XM_011538370.2 | 1588 | Silent Mutation | GCA,GCG | A,A 138 | XP_011536672.1 | |
XM_011538371.2 | 1588 | Silent Mutation | GCA,GCG | A,A 138 | XP_011536673.1 |