Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATTTGCTCTTGAGTCCACAGGT[A/C]CCACCTCTCCAAGAACGAAACAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604753 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF268 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF268 - zinc finger protein 268 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165881.2 | 366 | Silent Mutation | GTA,GTC | V,V 12 | NP_001159353.1 | |
NM_001165882.2 | 366 | UTR 5 | NP_001159354.1 | |||
NM_001165883.1 | 366 | Silent Mutation | GTA,GTC | V,V 12 | NP_001159355.1 | |
NM_001165884.2 | 366 | Intron | NP_001159356.2 | |||
NM_001165885.1 | 366 | Silent Mutation | GTA,GTC | V,V 12 | NP_001159357.1 | |
NM_001165886.1 | 366 | Intron | NP_001159358.1 | |||
NM_001165887.1 | 366 | Intron | NP_001159359.1 | |||
NM_003415.2 | 366 | Silent Mutation | GTA,GTC | V,V 12 | NP_003406.1 | |
NM_152943.2 | 366 | Silent Mutation | GTA,GTC | V,V 12 | NP_694422.2 |