Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACACATTGTAAGACTTTATTCAG[A/G]GTCTTACAGCAAAATAGTAGGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf73 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf73 - chromosome 12 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDG - thymine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003211.4 | Intron | NP_003202.3 | ||||
XM_005269125.1 | Intron | XP_005269182.1 | ||||
XM_011538714.1 | Intron | XP_011537016.1 | ||||
XM_011538715.1 | Intron | XP_011537017.1 |