Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCATCCAGCAGGGAGGGGACGA[C/T]GTGCAGCAGCTGTGTGAGGCGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602381 MIM: 616496 MIM: 601512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAB2 - NGFI-A binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005967.3 | 582 | Silent Mutation | GAC,GAT | D,D 68 | NP_005958.1 | |
XM_005268894.2 | 582 | Silent Mutation | GAC,GAT | D,D 68 | XP_005268951.1 |
NEMP1 - nuclear envelope integral membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT6 - signal transducer and activator of transcription 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |