Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTCAACAGCCAGATCTCATTG[C/T]GCCGTCCGTAGGGCTTCATGGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605826 | ||||||||||||||||||||
Literature Links: |
FAM234B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM234B - family with sequence similarity 234 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020853.1 | 737 | Intron | NP_065904.1 | |||
XM_017019706.1 | 737 | Intron | XP_016875195.1 |
HEBP1 - heme binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015987.4 | 737 | Missense Mutation | CAC,CGC | H,R 181 | NP_057071.2 |
LOC100506314 - uncharacterized LOC100506314 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |