Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGAGGGGCTTCCCCCCGCGGTT[A/G]TTCTCACCCACATCCAGGCATTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605148 | ||||||||||||||||||||
Literature Links: |
GALNT6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT6 - polypeptide N-acetylgalactosaminyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007210.3 | 2081 | Silent Mutation | AAC,AAT | N,N 515 | NP_009141.2 | |
XM_005268607.1 | 2081 | Silent Mutation | AAC,AAT | N,N 515 | XP_005268664.1 | |
XM_006719214.2 | 2081 | Silent Mutation | AAC,AAT | N,N 515 | XP_006719277.1 | |
XM_011537819.2 | 2081 | Silent Mutation | AAC,AAT | N,N 515 | XP_011536121.1 | |
XM_011537822.2 | 2081 | Silent Mutation | AAC,AAT | N,N 515 | XP_011536124.1 | |
XM_017018744.1 | 2081 | Silent Mutation | AAC,AAT | N,N 515 | XP_016874233.1 | |
XM_017018745.1 | 2081 | Silent Mutation | AAC,AAT | N,N 515 | XP_016874234.1 |