Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCACCACTGAGACCAGTGCCCAT[C/G]GAGCTTCTTCCTGCACATTTAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601933 MIM: 616929 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRY1 - cryptochrome circadian clock 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004075.4 | 2564 | Silent Mutation | TCC,TCG | S,S 556 | NP_004066.1 | |
XM_017018832.1 | 2564 | Silent Mutation | TCC,TCG | S,S 528 | XP_016874321.1 |
MTERF2 - mitochondrial transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |