Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTGGTAGGTGTGAGGACATTCC[A/G]AAACACGGCCATCCACATTCTGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 136351 | ||||||||||||||||||||
Literature Links: |
FLT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLT3 - fms related tyrosine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004119.2 | 2872 | Missense Mutation | TCG,TTG | S,L 963 | NP_004110.2 | |
XM_011535015.2 | 2872 | Missense Mutation | TCG,TTG | S,L 944 | XP_011533317.1 | |
XM_011535017.2 | 2872 | Missense Mutation | TCG,TTG | S,L 788 | XP_011533319.1 | |
XM_011535018.2 | 2872 | Missense Mutation | TCG,TTG | S,L 788 | XP_011533320.1 | |
XM_017020486.1 | 2872 | Missense Mutation | TCG,TTG | S,L 891 | XP_016875975.1 | |
XM_017020487.1 | 2872 | Missense Mutation | TCG,TTG | S,L 788 | XP_016875976.1 | |
XM_017020488.1 | 2872 | Missense Mutation | TCG,TTG | S,L 670 | XP_016875977.1 | |
XM_017020489.1 | 2872 | Missense Mutation | TCG,TTG | S,L 664 | XP_016875978.1 |