Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACCCAGGAAGGCCTACTGGCT[A/G]TGCAGCTACTTGGACTCCCGTTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD10 - ankyrin repeat domain 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286721.1 | 1246 | Intron | NP_001273650.1 | |||
NM_017664.3 | 1246 | Missense Mutation | ACA,ATA | T,I 358 | NP_060134.2 |
LINC00346 - long intergenic non-protein coding RNA 346 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |