Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGCTGGATAGCTTTCCAGAGC[C/G]GAAGCTGAATGGACCTGATGGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PROSER1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PROSER1 - proline and serine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025138.4 | 3534 | Silent Mutation | TCC,TCG | S,S 909 | NP_079414.3 | |
XM_005266545.3 | 3534 | Silent Mutation | TCC,TCG | S,S 887 | XP_005266602.1 | |
XM_011535239.2 | 3534 | Silent Mutation | TCC,TCG | S,S 880 | XP_011533541.1 |