Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGAAGAGTCCTCACCACTCTGC[A/G]TGTGGTTGATGAGATCCTTTTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDH8 - protocadherin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002590.3 | 3035 | Missense Mutation | ACG,ATG | T,M 944 | NP_002581.2 | |
NM_032949.2 | 3035 | Missense Mutation | ACG,ATG | T,M 847 | NP_116567.1 |