Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTGCTGGAAAAAGTACAGATGT[A/G]TAAGATGGGAAGCAACCTGGACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615794 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNDC3A - fibronectin type III domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079673.1 | Intron | NP_001073141.1 | ||||
NM_001278438.1 | Intron | NP_001265367.1 | ||||
NM_014923.4 | Intron | NP_055738.3 | ||||
XM_011534997.2 | Intron | XP_011533299.1 | ||||
XM_017020440.1 | Intron | XP_016875929.1 | ||||
XM_017020441.1 | Intron | XP_016875930.1 | ||||
XM_017020442.1 | Intron | XP_016875931.1 | ||||
XM_017020443.1 | Intron | XP_016875932.1 |