Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCAGAATGAAATAGAAACATTTT[C/T]GAGAAGTCTGCCAGCACTGAGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613408 MIM: 613409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC122 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC122 - coiled-coil domain containing 122 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LACC1 - laccase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128303.1 | 824 | Missense Mutation | TCG,TTG | S,L 168 | NP_001121775.1 | |
NM_153218.2 | 824 | Missense Mutation | TCG,TTG | S,L 168 | NP_694950.2 | |
XM_005266261.3 | 824 | Missense Mutation | TCG,TTG | S,L 168 | XP_005266318.1 | |
XM_006719766.3 | 824 | Missense Mutation | TCG,TTG | S,L 168 | XP_006719829.1 | |
XM_006719767.3 | 824 | Missense Mutation | TCG,TTG | S,L 168 | XP_006719830.1 | |
XM_011534933.2 | 824 | Missense Mutation | TCG,TTG | S,L 168 | XP_011533235.1 | |
XM_011534934.2 | 824 | Missense Mutation | TCG,TTG | S,L 168 | XP_011533236.1 | |
XM_011534935.2 | 824 | Missense Mutation | TCG,TTG | S,L 168 | XP_011533237.1 | |
XM_017020394.1 | 824 | Missense Mutation | TCG,TTG | S,L 168 | XP_016875883.1 |